siRNA Details
| Gene Symbol: Pradc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028081.1 | 1 | 172 | |||
| GenBank mRNA | BG088874.2 | - | 1 | 147 | ||
| CJ075544.1 | - | 1 | 172 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pradc1| Gene Symbol: Pradc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028081.1 | 1 | 172 | |||
| GenBank mRNA | BG088874.2 | - | 1 | 147 | ||
| CJ075544.1 | - | 1 | 172 | |||