siRNA Details
Gene Symbol: 9430083A17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029463.1 | 1 | 2578 | |||
GenBank mRNA | AK157883.1 | - | 1 | 2571 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 9430083A17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029463.1 | 1 | 2578 | |||
GenBank mRNA | AK157883.1 | - | 1 | 2571 |