siRNA Details
| Gene Symbol: Mir22hg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_030711.1 | 3 | 1075 | |||
| GenBank mRNA | AK008813.1 | - | LIT3441 2210403K04Rik |
3 | 967 | |
| AK018628.1 | - | 3 | 985 | |||

Mir22hg
Genomic Map