siRNA Details
| Gene Symbol: Snora7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028546.1 | 1 | 102 | |||
| GenBank mRNA | AF357398.1 | - | 1 | 102 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DT918153.1 | - | 1 | 347 | Chr. 6 - 115807111 - 115808348 [-] on Build GRCm38 | |

Snora7a
Genomic Map