siRNA Details
| Gene Symbol: 4931409K22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033188.1 | 6 | 1190 | |||
| GenBank mRNA | AK029839.1 | - | 6 | 1132 | ||
| BC137999.1 | - | 3 | 421 | 8861018 | ||
| BC145792.1 | - | 3 | 421 | 40131095 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4931409K22Rik| Gene Symbol: 4931409K22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033188.1 | 6 | 1190 | |||
| GenBank mRNA | AK029839.1 | - | 6 | 1132 | ||
| BC137999.1 | - | 3 | 421 | 8861018 | ||
| BC145792.1 | - | 3 | 421 | 40131095 | ||