siRNA Details
| Gene Symbol: Wbscr25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026907.1 | 4 | 1863 | |||
| GenBank mRNA | AK017044.1 | - | 4 | 1475 | ||
| AY081954.2 | - | 4 | 1863 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wbscr25| Gene Symbol: Wbscr25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026907.1 | 4 | 1863 | |||
| GenBank mRNA | AK017044.1 | - | 4 | 1475 | ||
| AY081954.2 | - | 4 | 1863 | |||