siRNA Details
| Gene Symbol: Wbscr25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026907.1 | 1 | 997 | |||
| GenBank mRNA | AK017044.1 | - | 1 | 610 | ||
| AK017130.1 | - | 1 | 154 | |||
| AY081954.2 | - | 1 | 997 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wbscr25| Gene Symbol: Wbscr25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026907.1 | 1 | 997 | |||
| GenBank mRNA | AK017044.1 | - | 1 | 610 | ||
| AK017130.1 | - | 1 | 154 | |||
| AY081954.2 | - | 1 | 997 | |||