siRNA Details
| Gene Symbol: Jpx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK137473.1 | - | 3 | 1582 | ||
| AK148110.1 | - | 3 | 1631 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Jpx| Gene Symbol: Jpx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK137473.1 | - | 3 | 1582 | ||
| AK148110.1 | - | 3 | 1631 | |||