siRNA Details
| Gene Symbol: 1700018A04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029439.1 | 5 | 533 | |||
| GenBank mRNA | AK006076.1 | - | 5 | 526 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY320487.1 | - | 5 | 348 | Chr. 13 - 31567309 - 31625153 [-] on Build GRCm38 | |

1700018A04Rik
Genomic Map