siRNA Details
Gene Symbol: 1700018A04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029439.1 | 6 | 698 | |||
GenBank mRNA | AK006076.1 | - | 6 | 691 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700018A04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029439.1 | 6 | 698 | |||
GenBank mRNA | AK006076.1 | - | 6 | 691 |