siRNA Details
| Gene Symbol: 1700018A04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029439.1 | 2 | 245 | |||
| GenBank mRNA | AK006076.1 | - | 2 | 238 | ||
| AK016091.1 | - | 2 | 273 | |||
| AK017122.1 | - | 2 | 246 | |||
| AK142058.1 | - | 767 | ||||
| BC049749.1 | - | 264 | ||||

1700018A04Rik
Genomic Map