siRNA Details
| Gene Symbol: 1700018A04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029439.1 | 3 | 340 | |||
| GenBank mRNA | AK006076.1 | - | 3 | 333 | ||
| AK016091.1 | - | 3 | 368 | |||
| AK142058.1 | - | 1146 | ||||
| BC049749.1 | - | 359 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700018A04Rik| Gene Symbol: 1700018A04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029439.1 | 3 | 340 | |||
| GenBank mRNA | AK006076.1 | - | 3 | 333 | ||
| AK016091.1 | - | 3 | 368 | |||
| AK142058.1 | - | 1146 | ||||
| BC049749.1 | - | 359 | ||||