siRNA Details
| Gene Symbol: Gm5166 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027707.1 | 8 | 864 | |||
| GenBank mRNA | BC049655.1 | - | 8 | 864 | ||
| Gene Symbol: 1700031F05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006578.1 | - | 7 | 1336 | ||

Gm5166
Genomic Map