siRNA Details
| Gene Symbol: Gm5166 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027707.1 | 2, 3 | 228 | |||
| GenBank mRNA | BC049655.1 | - | 2, 3 | 228 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CA463909.1 | - | 2, 3 | 231 | Chr. X - 102791193 - 102796724 [+] on Build GRCm38 | |

Gm5166
Genomic Map