siRNA Details
| Gene Symbol: Gm5166 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027707.1 | 7 | 804 | |||
| GenBank mRNA | BC049655.1 | - | 7 | 804 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5166| Gene Symbol: Gm5166 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027707.1 | 7 | 804 | |||
| GenBank mRNA | BC049655.1 | - | 7 | 804 | ||