siRNA Details
| Gene Symbol: Fxyd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028499.1 | 1 | 50 | |||
| GenBank mRNA | CA491597.1 | - | 1 | 84 | ||
| CB600958.1 | - | 1 | 53 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fxyd2| Gene Symbol: Fxyd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028499.1 | 1 | 50 | |||
| GenBank mRNA | CA491597.1 | - | 1 | 84 | ||
| CB600958.1 | - | 1 | 53 | |||