siRNA Details
| Gene Symbol: Atp2a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027838.1 | 20 | 4827 | |||
| GenBank mRNA | AK081191.1 | - | 1 | 464 | ||
| AK134659.1 | - | 1 | 996 | |||
| AK220538.1 | - | 20 | 4776 | |||
| BC054531.1 | - | 20 | 4486 | 6406916 | ||
| BC054748.1 | - | 20 | 4486 | 6406916 | ||

Atp2a2
Genomic Map