siRNA Details
| Gene Symbol: Gm8883 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027658.1 | 3 | 720 | |||
| NR_027659.1 | 3 | 400 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm8883| Gene Symbol: Gm8883 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027658.1 | 3 | 720 | |||
| NR_027659.1 | 3 | 400 | ||||