siRNA Details
| Gene Symbol: Gm8883 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027658.1 | 2 | 468 | |||
| NR_027659.1 | 2 | 148 | ||||
| GenBank mRNA | BX527230.1 | - | 2 | 487 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm8883| Gene Symbol: Gm8883 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027658.1 | 2 | 468 | |||
| NR_027659.1 | 2 | 148 | ||||
| GenBank mRNA | BX527230.1 | - | 2 | 487 | ||