siRNA Details
| Gene Symbol: 1700001L05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027980.1 | 4 | 4917 | |||
| GenBank mRNA | DQ699522.1 | - | 1 | 20 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700001L05Rik| Gene Symbol: 1700001L05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027980.1 | 4 | 4917 | |||
| GenBank mRNA | DQ699522.1 | - | 1 | 20 | ||