siRNA Details
| Gene Symbol: 1700001L05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027980.1 | 2 | 709 | |||
| GenBank mRNA | AK005608.1 | - | 2 | 546 | ||
| AK132681.1 | - | 2 | 690 | |||
| BC099567.1 | - | 2 | 697 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700001L05Rik| Gene Symbol: 1700001L05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027980.1 | 2 | 709 | |||
| GenBank mRNA | AK005608.1 | - | 2 | 546 | ||
| AK132681.1 | - | 2 | 690 | |||
| BC099567.1 | - | 2 | 697 | |||