siRNA Details
| Gene Symbol: Etohd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015349.2 | 1 | 19 | |||
| GenBank mRNA | AK017511.1 | - | 1 | 23 | ||
| AK035969.1 | - | 1 | 20 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB628651.1 | - | 1 | 17 | Chr. 13 - 59769968 - 59771211 [+] on Build GRCm38 | |

Etohd2
Genomic Map