siRNA Details
| Gene Symbol: Etohd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015349.2 | 2 | 756 | |||
| GenBank mRNA | AK017511.1 | - | 2 | 547 | ||
| AK035969.1 | - | 2 | 757 | |||
| AK042838.1 | - | 2 | 700 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Etohd2| Gene Symbol: Etohd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015349.2 | 2 | 756 | |||
| GenBank mRNA | AK017511.1 | - | 2 | 547 | ||
| AK035969.1 | - | 2 | 757 | |||
| AK042838.1 | - | 2 | 700 | |||