siRNA Details
| Gene Symbol: 2900005J15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027851.1 | 1 | 1334 | |||
| GenBank mRNA | AK015504.1 | - | 851 | |||
| AK076604.1 | - | 1 | 1328 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | NM_026059.1 | - | 1328 | Chr. - - [-] on Build | ||

2900005J15Rik
Genomic Map