siRNA Details
| Gene Symbol: 2900005J15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027851.1 | 1 | 891 | |||
| GenBank mRNA | AK013483.1 | - | 1 | 892 | ||
| AK015504.1 | - | 408 | ||||
| BC094288.1 | - | 1 | 926 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2900005J15Rik| Gene Symbol: 2900005J15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027851.1 | 1 | 891 | |||
| GenBank mRNA | AK013483.1 | - | 1 | 892 | ||
| AK015504.1 | - | 408 | ||||
| BC094288.1 | - | 1 | 926 | |||