siRNA Details
| Gene Symbol: AI506816 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015554.2 | 5 | 2294 | |||
| GenBank mRNA | AK088877.1 | - | 3254 | |||
| AK089743.1 | - | 5 | 2335 | |||
| AK157274.1 | - | 5 | 2201 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AI506816| Gene Symbol: AI506816 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015554.2 | 5 | 2294 | |||
| GenBank mRNA | AK088877.1 | - | 3254 | |||
| AK089743.1 | - | 5 | 2335 | |||
| AK157274.1 | - | 5 | 2201 | |||