siRNA Details
| Gene Symbol: AI506816 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK044688.1 | - | 425 | |||
| AK088877.1 | - | 349 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AI506816| Gene Symbol: AI506816 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK044688.1 | - | 425 | |||
| AK088877.1 | - | 349 | ||||