siRNA Details
| Gene Symbol: 1110002L01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030694.1 | 1 | 212 | |||
| GenBank mRNA | AK031200.1 | - | 1 | 109 | ||
| AK035515.1 | - | 1 | 109 | |||
| BY152001.1 | - | 1 | 213 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1110002L01Rik| Gene Symbol: 1110002L01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030694.1 | 1 | 212 | |||
| GenBank mRNA | AK031200.1 | - | 1 | 109 | ||
| AK035515.1 | - | 1 | 109 | |||
| BY152001.1 | - | 1 | 213 | |||