siRNA Details
| Gene Symbol: 1110002L01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030694.1 | 3 | 1914 | |||
| GenBank mRNA | AK003311.1 | - | 1 | 54 | ||
| AK035515.1 | - | 3 | 1811 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1110002L01Rik| Gene Symbol: 1110002L01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030694.1 | 3 | 1914 | |||
| GenBank mRNA | AK003311.1 | - | 1 | 54 | ||
| AK035515.1 | - | 3 | 1811 | |||