siRNA Details
| Gene Symbol: Nnt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK085903.1 | - | 15 | 3037 | ||
| AK192828.1 | - | 1 | 258 | |||
| BC008518.1 | - | 17 | 4105 | 3601702 | ||
| CJ192946.1 | - | 1 | 436 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nnt| Gene Symbol: Nnt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK085903.1 | - | 15 | 3037 | ||
| AK192828.1 | - | 1 | 258 | |||
| BC008518.1 | - | 17 | 4105 | 3601702 | ||
| CJ192946.1 | - | 1 | 436 | |||