siRNA Details
| Gene Symbol: Hoxd3os1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027899.1 | 3 | 573 | |||
| GenBank mRNA | AK007148.1 | - | 3 | 441 | ||
| AK054396.1 | - | 2 | 574 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hoxd3os1| Gene Symbol: Hoxd3os1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027899.1 | 3 | 573 | |||
| GenBank mRNA | AK007148.1 | - | 3 | 441 | ||
| AK054396.1 | - | 2 | 574 | |||