siRNA Details
| Gene Symbol: Hoxd3os1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027899.1 | 2 | 506 | |||
| GenBank mRNA | AK006878.1 | - | 2 | 359 | ||
| AK007148.1 | - | 2 | 374 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hoxd3os1| Gene Symbol: Hoxd3os1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027899.1 | 2 | 506 | |||
| GenBank mRNA | AK006878.1 | - | 2 | 359 | ||
| AK007148.1 | - | 2 | 374 | |||