siRNA Details
| Gene Symbol: Ufd1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028403.1 | 2, 3 | 228 | |||
| GenBank mRNA | CJ053815.1 | - | 2, 3 | 223 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ufd1l| Gene Symbol: Ufd1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028403.1 | 2, 3 | 228 | |||
| GenBank mRNA | CJ053815.1 | - | 2, 3 | 223 | ||