siRNA Details
| Gene Symbol: Gm5126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026596.1 | 1 | 2110 | |||
| GenBank mRNA | AK077703.1 | - | 1 | 2111 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5126| Gene Symbol: Gm5126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026596.1 | 1 | 2110 | |||
| GenBank mRNA | AK077703.1 | - | 1 | 2111 | ||