siRNA Details
| Gene Symbol: 3110021A11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030776.1 | 1 | 627 | |||
| GenBank mRNA | AK014066.1 | - | 1 | 628 | ||
| AK014105.1 | - | 1 | 610 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

3110021A11Rik| Gene Symbol: 3110021A11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030776.1 | 1 | 627 | |||
| GenBank mRNA | AK014066.1 | - | 1 | 628 | ||
| AK014105.1 | - | 1 | 610 | |||