siRNA Details
| Gene Symbol: 3110021A11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030776.1 | 1 | 481 | |||
| GenBank mRNA | AK014066.1 | - | 1 | 482 | ||
| AK014105.1 | - | 1 | 464 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY655184.1 | - | 1 | 74 | Chr. 6 - 119848600 - 119849014 [+] on Build GRCm38 | |

3110021A11Rik
Genomic Map