siRNA Details
| Gene Symbol: 3110039M20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026733.1 | 3 | 739 | |||
| GenBank mRNA | AK014152.1 | - | 2 | 432 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY713129.1 | - | 2 | 432 | Chr. 12 - 49390424 - 49407331 [+] on Build GRCm38 | |

3110039M20Rik
Genomic Map