siRNA Details
| Gene Symbol: 3110039M20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026733.1 | 3 | 656 | |||
| GenBank mRNA | AK014152.1 | - | 2 | 349 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

3110039M20Rik| Gene Symbol: 3110039M20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026733.1 | 3 | 656 | |||
| GenBank mRNA | AK014152.1 | - | 2 | 349 | ||