siRNA Details
| Gene Symbol: 3110039M20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026733.1 | 3 | 487 | |||
| GenBank mRNA | AK014151.1 | - | 3 | 252 | ||
| AK014152.1 | - | 2 | 180 | |||
| AK019435.1 | - | 3 | 252 | |||
| AK043991.1 | - | 2 | 229 | |||
| CF744769.1 | - | 3 | 487 | |||

3110039M20Rik
Genomic Map