siRNA Details
| Gene Symbol: 3110070M22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027974.1 | 1 | 946 | |||
| GenBank mRNA | AK005613.1 | - | 1 | 938 | ||
| AK014250.1 | - | 1 | 946 | |||
| BC050963.1 | - | 1 | 946 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

3110070M22Rik| Gene Symbol: 3110070M22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027974.1 | 1 | 946 | |||
| GenBank mRNA | AK005613.1 | - | 1 | 938 | ||
| AK014250.1 | - | 1 | 946 | |||
| BC050963.1 | - | 1 | 946 | |||