siRNA Details
Gene Symbol: 3110070M22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027974.1 | 1 | 780 | |||
GenBank mRNA | AK005613.1 | - | 1 | 772 | ||
AK014250.1 | - | 1 | 780 | |||
BC050963.1 | - | 1 | 780 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 3110070M22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027974.1 | 1 | 780 | |||
GenBank mRNA | AK005613.1 | - | 1 | 772 | ||
AK014250.1 | - | 1 | 780 | |||
BC050963.1 | - | 1 | 780 |