siRNA Details
| Gene Symbol: 3110070M22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027974.1 | 1 | 489 | |||
| GenBank mRNA | AK005613.1 | - | 1 | 481 | ||
| AK014250.1 | - | 1 | 489 | |||
| BC050963.1 | - | 1 | 489 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BU935752.1 | - | 1 | 15 | Chr. 13 - 119487224 - 119487721 [-] on Build GRCm38 | |

3110070M22Rik
Genomic Map