siRNA Details
| Gene Symbol: Vmn2r-ps11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003962.1 | 1 | 55 | |||
| GenBank mRNA | AK029734.1 | - | 1 | 56 | ||
| BB615497.1 | - | 1 | 56 | |||
| Gene Symbol: Vmn2r7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK030224.1 | - | 77 | |||

Vmn2r-ps11
Genomic Map