siRNA Details
| Gene Symbol: Vmn2r-ps11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003962.1 | 8, 9 | 2032 | |||
| GenBank mRNA | AK029734.1 | - | 8, 9 | 2033 | ||
| AV266491.2 | - | 3, 4 | 448 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn2r-ps11| Gene Symbol: Vmn2r-ps11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003962.1 | 8, 9 | 2032 | |||
| GenBank mRNA | AK029734.1 | - | 8, 9 | 2033 | ||
| AV266491.2 | - | 3, 4 | 448 | |||