siRNA Details
| Gene Symbol: 2700097O09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030764.1 | 4, 5 | 595 | |||
| GenBank mRNA | CX231570.1 | - | 4, 5 | 661 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2700097O09Rik| Gene Symbol: 2700097O09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030764.1 | 4, 5 | 595 | |||
| GenBank mRNA | CX231570.1 | - | 4, 5 | 661 | ||