siRNA Details
| Gene Symbol: 5430416O09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033355.1 | 2 | 1984 | |||
| GenBank mRNA | AK030334.1 | - | 2 | 1985 | ||
| BC147344.1 | - | 1 | 1516 | 9056418 | ||
| BC147345.1 | - | 1 | 1516 | 9056411 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

5430416O09Rik| Gene Symbol: 5430416O09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033355.1 | 2 | 1984 | |||
| GenBank mRNA | AK030334.1 | - | 2 | 1985 | ||
| BC147344.1 | - | 1 | 1516 | 9056418 | ||
| BC147345.1 | - | 1 | 1516 | 9056411 | ||