siRNA Details
| Gene Symbol: Rmst | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028262.1 | 13 | 2307 | |||
| GenBank mRNA | AK086758.1 | - | 13 | 2301 | ||
| AK136927.1 | - | 9 | 2656 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rmst| Gene Symbol: Rmst | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028262.1 | 13 | 2307 | |||
| GenBank mRNA | AK086758.1 | - | 13 | 2301 | ||
| AK136927.1 | - | 9 | 2656 | |||