siRNA Details
| Gene Symbol: Rmst | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028262.1 | 12 | 1391 | |||
| GenBank mRNA | AK086758.1 | - | 12 | 1384 | ||
| AK136927.1 | - | 7 | 2014 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BQ572455.1 | - | 8 | 731 | Chr. 10 - 92083887 - 92164663 [-] on Build GRCm38 | |

Rmst
Genomic Map