siRNA Details
| Gene Symbol: 1700018L02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028360.1 | 1 | 1145 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK006210.1 | - | 1094 | Chr. - - [-] on Build | ||

1700018L02Rik
Genomic Map