siRNA Details
| Gene Symbol: 1700047M11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015458.1 | 2 | 529 | |||
| GenBank mRNA | AK006718.1 | - | 2 | 313 | ||
| AK019698.1 | - | 2 | 308 | |||
| AK047804.1 | - | 2290 | ||||
| AV345418.1 | - | 1 | 80 | |||
| BC048591.1 | - | 2 | 529 | |||

1700047M11Rik
Genomic Map